Learn More
Caspase 4 Mouse anti-Human, Unconjugated, Clone: 1D1E12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67398-1-IG-20UL
Description
The Caspase 4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Caspase 4. This antibody recognizes human antigen. The Caspase 4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Caspase 4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D1E12 | |
| Unconjugated | |
| 20 μL | |
| 837 | |
| IgG1 |