missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caspase-4 Monoclonal antibody specifically detects Caspase-4 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Caspase-4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4A2 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | apoptotic cysteine protease Mih1/TX, CASP-4, caspase 4, apoptosis-related cysteine peptidase, caspase 4, apoptosis-related cysteine protease, caspase-4, EC 3.4.22, EC 3.4.22.57, ICE(rel)II, ICE(rel)-II, ICEREL-II, ICH2, ICH-2, Mih1/TX, Protease ICH-2, Protease TX, TX |
| Host Species | Mouse |
| Immunogen | Recombinant protein fragment corresponding to amino acids 81-270 of human Caspase-4 (NP_150649) produced in E.coli. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?