missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRDG1/STAP-1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06491F
This item is not returnable.
View return policy
Description
BRDG1/STAP-1 Polyclonal antibody specifically detects BRDG1/STAP-1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| BRDG1/STAP-1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| BCR downstream-signaling protein 1, BRDG1STAP-1BCR downstream signaling 1, Docking protein BRDG1, signal transducing adaptor family member 1, signal-transducing adaptor protein 1, signal-transducing adaptor protein-1, Stem cell adaptor protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human BRDG1/STAP-1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 26228 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction