missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Rabbit anti-Human, Mouse, Rat, Clone: 1U1D9, Novus Biologicals™
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human, Mouse, Rat samples. It is validated for Western Blot, KnockDown
Specifications
Specifications
| Antigen | Caspase-3 |
| Applications | Western Blot, KnockDown |
| Classification | Monoclonal |
| Clone | 1U1D9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, Knockout Validated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 29-175 of human Caspase-3 (P42574). SGISLDNSYKMDYPEMGLCIIINNKNFHKSTGMTSRSGTDVDAANLRETFRNLKYEVRNKNDLTREEIVELMRDVSKEDHSKRSSFVCVLLSHGEEGIIFGTNGPVDLKKITNFFRGDRCRSLTGKPKLFIIQACRGTELDCGIETD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?