missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caspase 3 p17 Monoclonal Antibody (31A1067)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547699
This item is not returnable.
View return policy
Description
The antibody detects pro Caspase-3 at 32 kDa* and the large subunit of the cleaved form of Caspase-3 at 14-21 kDa. The antibody can be used for western blot, immunohistochemistry and immuno-electron microscopy.
CASP3 (Caspase3 Apoptosis-Related Cysteine Protease), also known as YAMA, CPP32 or APOPAIN, is a caspase protein that interacts with caspase 8 and caspase 9. It is encoded by the CASP3 gene. The CASP3 protein is a member of the cysteine-aspartic acid protease (caspase) family. Tiso et al. (1996) used radiation hybrid mapping to localize the CPP32 gene to human chromosome 4q33-q35.1. Fernandes-Alnemri et al. (1994) found that overexpression of CPP32 in insect cells induced apoptosis. Coexpression of the 2 CPP32 subunits in insect cells also resulted in apoptosis. Tewari et al. (1995) showed that purified human ICE cleaved the Yama proenzyme into a proteolytically active form and that activated Yama cleaved PARP into the 85-kD apoptotic form.
Specifications
| Caspase 3 p17 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P42574, P55213, P70677 | |
| CASP3 | |
| Clone 31A1067 was developed against full-length recombinant human Caspase-3. The antibody recognizes an epitope in the large domain subunit (p17) of Caspase-3, and it will recognize pro Caspase-3 and the p17 cleavage fragment. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 31A1067 | |
| Unconjugated | |
| CASP3 | |
| A830040C14Rik; AC-3; Apopain; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; CC3; Cleaved Caspase 3; Cpp32; CPP-32; CPP32beta; cysteine protease CPP32; IRP; LICE; mldy; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 12367, 25402, 836 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction