missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody (S02-7C2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20021-50ul
This item is not returnable.
View return policy
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Caspase-3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry 1:20, Immunocytochemistry/ Immunofluorescence 10 μg/ml, Immunohistochemistry-Paraffin | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| A synthetic peptide of human Caspase-3 (Uniprot # P42574) | |
| 50 μg | |
| Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction | |
| 836 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S02-7C2 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction