Learn More
Eg5 Antibody (1G11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26182-100ul
 View more versions of this product
Product Code. 30039629
  • £406.00 / 100µL
Description
Eg5 Monoclonal antibody specifically detects Eg5 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Eg5
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
1G11
Western Blot 1:500-1:5000, ELISA
Eg5, HKSP, kinesin family member 11, kinesin-like 1, Kinesin-like protein 1, kinesin-like protein KIF11, Kinesin-like spindle protein HKSP, Kinesin-related motor protein Eg5, KNSL1TRIP-5, thyroid receptor interacting protein 5, Thyroid receptor-interacting protein 5, TR-interacting protein 5, TRIP5EG5
A synthesized peptide derived from Human Eg5 [UniProt P52732]
100 μL
Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Phospho Specific, Signal Transduction, Stem Cell Markers
3832
Store at -20 to -70C. Avoid freeze-thaw cycles.