missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-3 Antibody (14C1), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Caspase-3 |
|---|---|
| Clone | 14C1 |
| Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043623
|
Novus Biologicals
NBP3-26698-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043595
|
Novus Biologicals
NBP3-26698-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-3 Monoclonal antibody specifically detects Caspase-3 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Caspase-3 | |
| Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Apopain, apoptosis-related cysteine protease, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
| A synthesized peptide derived from Human Caspase-3 [UniProt P42574] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 14C1 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Cancer, Caspases, Cell Biology, Lipid and Metabolism, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 836 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title