missing translation for 'onlineSavingsMsg'
Learn More

Caspase-2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18426126 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426126 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426126 Supplier Novus Biologicals Supplier No. NBP226612DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase-2 Polyclonal antibody specifically detects Caspase-2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from the amino-terminus of human Caspase-2. The immunogen is located within the first 50 amino acids of Caspase-2. Amino Acid Squence: CGMHPHHQETLKKNRV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 835
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.