missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-2 Antibody (7E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26464-100ul
This item is not returnable.
View return policy
Description
Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Caspase-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 7E5 | |
| Western Blot 1:500-1:5000, ELISA | |
| caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57 | |
| A synthesized peptide derived from Human Caspase-2 [UniProt P42575] | |
| 100 μL | |
| Apoptosis, Cancer, Caspases | |
| 835 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction