missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caspase-14 Polyclonal antibody specifically detects Caspase-14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin) |
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Research Discipline | Apoptosis, Cancer, Caspases |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?