missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Caspase-14 Polyclonal antibody specifically detects Caspase-14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Formulation | 50mM Sodium Borate |
| Gene Alias | apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE |
| Host Species | Rabbit |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?