missing translation for 'onlineSavingsMsg'
Learn More

Caspase-14 Antibody (05), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-05778-100ul

 View more versions of this product

Product Code. 18649336

  • £248.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Caspase-14 Monoclonal antibody specifically detects Caspase-14 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Caspase-14
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA, Immunofluorescence
05
Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:1000
apoptosis-related cysteine protease, CASP-14, caspase 14, apoptosis-related cysteine peptidase, caspase 14, apoptosis-related cysteine protease, caspase-14, EC 3.4.22.-, MGC119078, MGC119079, MICE
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Caspase-14 (Accession#: NP_036246.1; Ser 2-Gln 242).
100 μg
Apoptosis, Cancer, Caspases
23581
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.