missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Caspase 13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848162
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848162 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848162 Supplier Invitrogen™ Supplier No. PA519949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~43kDa. A suggested positive control is HL60 cell lysate. PA5-19949 can be used with blocking peptide PEP-0074. The immunogen is located within amino acids 80 - 130 of Caspase-13.

Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death is finally caused by members of the caspase family of proteases and caspase activated DNases. A novel member in the caspase family was recently identified and designated ERICE (for Evolutionarily Related Interleukin-1 beta Converting Enzyme) and caspase-13. Caspase-13 belongs to the ICE subfamily of caspases. Overexpression of caspase-13 induces apoptosis. Caspase-13 was activated by caspase-8, which is a key enzyme in death receptor induced apoptosis. Caspase-13 is expressed in a variety of human tissues and cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 13
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CASP13
Gene Accession No. O75601
Gene Alias apoptosis-related cysteine protease; Casp11; CASP-11; CASP13; CASP-13; CASP4; CASP-4; caspase 11; caspase 4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; caspase 4, isoform alpha, small subunit; Caspase-11; Caspase-13; Caspase-13 subunit 1; Caspase-13 subunit 2; caspase-13A; caspase-13B; caspase-4; Caspase-4 subunit 1; Caspase-4 subunit 2; Caspase-4 subunit p10; Caspase-4 subunit p20; Caspl; ERICE; Evolutionary related interleukin-1-beta-converting enzyme; Ich3; ich-3; MGC137679; Protease ICH-3
Gene Symbols CASP13
Host Species Rabbit
Immunogen A 15 amino acid peptide near the amino terminus of bovine Caspase-13.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338039, 8628
Target Species Bovine, Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.