missing translation for 'onlineSavingsMsg'
Learn More

Caspase-12 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18418444 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418444 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418444 Supplier Novus Biologicals Supplier No. NBP176801C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase-12 Polyclonal antibody specifically detects Caspase-12 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-12
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of murine Caspase-12. The immunogen is located within amino acids 90 - 140 of Caspase-12. Amino Acid Squence: CTPSSPSESKRKVEDDE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 100506742
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.