missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-12 Antibody (12G6), DyLight 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80101V
This item is not returnable.
View return policy
Description
Caspase-12 Monoclonal antibody specifically detects Caspase-12 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Caspase-12 | |
| Monoclonal | |
| DyLight 405 | |
| Apoptosis Related Cysteine Protease, CASP 12, casp12, CASP12P1, Caspase 12 pseudogene 1, Caspase12, UNQ9415 | |
| Recombinant mouse caspase-12 (p20 fragment) and synthetic peptide corresponding to aa 183-205 of mouse caspase-12. | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases | |
| 100506742 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA | |
| 12G6 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction