missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-11 Antibody (8A5), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80099PCP
This item is not returnable.
View return policy
Description
Caspase-11 Monoclonal antibody specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Caspase-11 | |
| Monoclonal | |
| PerCP | |
| ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl | |
| Recombinant mouse caspase-11 (p20 fragment). | |
| 0.1 mL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| 8A5 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| 12363 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction