missing translation for 'onlineSavingsMsg'
Learn More

Caspase-11 Antibody (4E11), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18400737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400737 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400737 Supplier Novus Biologicals Supplier No. NBP280100MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Caspase-11 Monoclonal antibody specifically detects Caspase-11 in Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-11
Applications Flow Cytometry
Classification Monoclonal
Clone 400000000000
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ich-3, CASP11, CASP-11, CASP4, CASP-4, caspase 4, apoptosis-related cysteine peptidase, Caspl
Host Species Rat
Immunogen Recombinant mouse caspase-11 (p20 fragment).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12363
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.