missing translation for 'onlineSavingsMsg'
Learn More

Caspase-10/FLICE2 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30734444 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734444 Supplier Novus Biologicals Supplier No. AF6864MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Caspase-10/FLICE2 Polyclonal antibody specifically detects Caspase-10/FLICE2 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-10/FLICE2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias Apoptotic protease Mch-4, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, MCH4ALPS2
Host Species Goat
Immunogen E. coli-derived recombinant human Pro Caspase-10, Met1-Pro228, Accession # Q92851
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Caspases
Primary or Secondary Primary
Gene ID (Entrez) 843
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.