missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 1/p20/p10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22915-1-AP-150UL
This item is not returnable.
View return policy
Description
22915-1-AP can recognize all five isoforms of caspase 1. It also recognizes p45 procaspase and the mature fragments p10 and p20.
Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.Specifications
| Caspase 1/p20/p10 | |
| Polyclonal | |
| Unconjugated | |
| CASP1 | |
| CASP 1, CASP1, Caspase 1, Caspase 1/p20/p10, Caspase1, ICE, IL 1 beta converting enzyme, IL 1BC, IL1BC, IL1BCE, Interleukin 1 beta convertase, p10, p20, P45 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12362, 25166, 834 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P29452, P29466, P43527 | |
| Casp1 | |
| Caspase 1/p20/p10 Fusion Protein Ag11143 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction