missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Caspase-1 Antibody Pair [Biotin]
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Specifications
Specifications
| Assay Range | 0.15 ng/mL to 10 ng/mL |
| Assay Sensitivity | 50 pg/mL |
| Conjugate | Biotin |
| Product Type | Sandwich ELISA |
| Sample Type | Cell Culture Supernatant |
| Specificity | Detects mouse Caspase-1 (p10 and p20 domain). Does not detect human Caspase-1. |
| For Use With (Application) | Sandwich ELISA |
| Gene ID (Entrez) | 834 |
| Gene Symbol | CASP1 |
| Quantity | 1 Set |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction