missing translation for 'onlineSavingsMsg'
Learn More

Caspase-1 Antibody, Novus Biologicals™

Product Code. 18422862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422862 25 μL 25µL
18461282 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18422862 Supplier Novus Biologicals Supplier No. NBP18768025ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase-1 Polyclonal antibody specifically detects Caspase-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias CASP1, CASP-1, CASP1 nirs variant 1, caspase 1, apoptosis-related cysteine peptidase, caspase-1, Caspase-1 subunit p10, Caspase-1 subunit p20, EC 3.4.22.36, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL1BC, IL-1BC, IL1BCE, IL1B-convertase, interleukin 1, beta, convertase, interleukin 1-B converting enzyme, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Host Species Rabbit
Immunogen Caspase-1 Antibody was developed against a recombinant protein corresponding to amino acids: PDILQLNAIFNMLNTKNCPSLKDKPKVIIIQACRGDSPGVVWFKDSVGVSGNLSLPTTEEFEDDAIKKAHIEKDFIAFCSSTPDNVSWRHPTMGSVFIGRLIEHMQEYACSCDVEEIFRKVRFSFEQPDGRAQMPTTERVTLTRCFYLFPGH
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology, Inflammation, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 834
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.