missing translation for 'onlineSavingsMsg'
Learn More

Caspase-1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18400336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400336 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400336 Supplier Novus Biologicals Supplier No. NBP176605MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Caspase-1 Polyclonal antibody specifically detects Caspase-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CASP1, CASP-1, CASP1 nirs variant 1, caspase 1, apoptosis-related cysteine peptidase, caspase-1, Caspase-1 subunit p10, Caspase-1 subunit p20, EC 3.4.22.36, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL1BC, IL-1BC, IL1BCE, IL1B-convertase, interleukin 1, beta, convertase, interleukin 1-B converting enzyme, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Host Species Rabbit
Immunogen Caspase-1 Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human Caspase-1. The immunogen is located within the last 50 amino acids of Caspase-1 (NP_001244047.1, NP_001244048.1, NP_150635.1, NP_150636.1). Amino Acid Squence: SFEQPDGRAQMPTTER
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Caspases, Cell Biology, Inflammation, Innate Immunity, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 834
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.