missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-1 Antibody (14F468) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 54 publications
£163.00 - £457.00
Specifications
| Dilution | Western Blot 0.5-2 ug/ml, Simple Western 1:50, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 30930743) |
|---|---|
| Gene Alias | CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18716323
|
Novus Biologicals
NB100-56565SS |
£163.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18758593
|
Novus Biologicals
NB100-56565 |
0.1 mg |
£457.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-1 Monoclonal specifically detects Caspase-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Western Blot 0.5-2 ug/ml, Simple Western 1:50, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 30930743) | |
| CASP-1, caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, caspase 1, apoptosis-related cysteine protease (interleukin 1, beta, caspase-1, EC 3.4.22, EC 3.4.22.36, ICEP45, IL-1 beta-converting enzyme, IL-1BC, IL1BC CASP1 nirs variant 1, IL1BCE, IL1B-convertase, interleukin 1-B converting enzyme, interleukin 1-beta convertase, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title