missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CASKIN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584882
This item is not returnable.
View return policy
Description
Immunogen sequence: APWAFSYLAG PPATPPDPPR PKRRSHSLSR PGPTEGDAEG EAEGPVGSTL GSYATLTRRP GRSALVRTSP SVTPTPARGT PRSQSFALR Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.
This gene encodes a large protein that contains six ankyrin repeats, as well as a Src homology 3 (SH3) domain and two sterile alpha motif (SAM) domains, which may be involved in protein-protein interactions. The C-terminal portion of this protein is proline-rich and contains a conserved region. A related protein interacts with calcium/calmodulin-dependent serine protein kinase (CASK). Alternative splicing results in multiple transcript variants.
Specifications
| CASKIN2 | |
| Polyclonal | |
| Unconjugated | |
| CASKIN2 | |
| 1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57513 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WXE0 | |
| CASKIN2 | |
| Recombinant protein corresponding to Human CASKIN2. Recombinant protein control fragment (Product #RP-108255). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction