missing translation for 'onlineSavingsMsg'
Learn More

CASIN, MedChemExpress

Product Code. 16483336
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483336

Brand: Medchem Express HY12874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CASIN is a selective GTPase Cdc42 inhibitor with an IC50 of 2 uM. CASIN can be used for the research of cancer.

CASIN is a selective GTPase Cdc42 inhibitor with an IC50 of 2 uM. CASIN can be used for the research of cancer.

Specifications

Chemical Name or Material CASIN
Physical Form Solid
CAS 425399-05-9
Color Light Nude
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C20H22N2O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 31 mg/mL (101.17 mM)
SMILES OCCNC1C(NC2=C3C=C(C4=CC=CC=C4)C=C2)=C3CCC1
Molecular Weight (g/mol) 306.4
Formula Weight 306.4
Percent Purity 95.28%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.