Learn More
Thermo Scientific™ Casein Sodium Salt
For research use
Specifications
| Formulation | Lyophilized powder |
|---|---|
| Source | Bovine milk |
| Regulatory Status | RUO |
| Product Type | Casein sodium salt |
| Recombinant | No |
Description
α-Casein can be an immunogen in young children, and anti-α-casein antibodies have strong cross-reactivity with insulin. This effect can explain the uncommon insulin autoimmunity, but it does not appear to progress to Type I diabetes.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Lyophilized powder | |
| RUO | |
| No |
| Bovine milk | |
| Casein sodium salt |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.