missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Casein Kinase 2α Protein, Active

Product Code. 17340184
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17340184

Brand: Eurofins DiscoverX 14445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, 6His-tagged recombinant full-length Casein Kinase 2α, human, for use in Kinase Assays.

Mammalian casein kinase 2α1 (CK2a1) is a highly conserved serine-threonine protein kinase that is constitutively expressed in all eukaryotic cell types including yeast, insects, and vertebrates. The enzyme can utilize GTP or ATP as a phosphate donor. Casein kinase activity is stimulated by mitogens in a cell type-specific manner during development and embryogenesis. Inhibition of CK2α1 may be useful in the treatment of human squamous cell carcinoma, adenocarcinoma of the lung, prostatic hyperplasia and lymphomas.

GenBank J02853.1

Specifications

Formulation mg/mL of enzyme in 20 mM MOPS pH 7.4, 1 mM EDTA, 50% glycerol, 1 mM benzamidine, 0.1% Triton X-100, 0.3% 2-mercaptoethanol. Liquid at -20°C.
Molecular Weight (g/mol) 48.7 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥69%
Protein Casein Kinase 2a, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.