missing translation for 'onlineSavingsMsg'
Learn More

Casein Kinase 2 beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-06515

Product Code. 18242820

  • £458.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Casein Kinase 2 beta Polyclonal specifically detects Casein Kinase 2 beta in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Casein Kinase 2 beta
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
casein kinase 2, beta polypeptide, Casein kinase II beta subunit, casein kinase II subunit beta, CK II beta, CK2B, CSK2B, G5A, MGC138222, MGC138224, phosvitin, Protein G5a
Rabbit
Affinity Purified
RUO
Primary
Human, Mouse
IgG
Western Blot, Immunoprecipitation
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
NP_001311.3
CSNK2B
The immunogen recognized by this antibody maps to a region between residue 165 and 215 of human casein kinase 2, beta using the numbering given in entry NP_001311.3 (GeneID 1460).
0.1 mL
Cancer, Cell Cycle and Replication, Wnt Signaling Pathway
1460
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only