missing translation for 'onlineSavingsMsg'
Learn More

Casein Kinase 2 alpha Antibody (S05-7F8), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £500.00

Specifications

Antigen Casein Kinase 2 alpha
Clone S05-7F8
Concentration 0.3 mg/mL
Applications Western Blot, Immunofluorescence, Immunoprecipitation, Gene Knock-Out
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18628713
View Documents
Novus Biologicals
NBP3-19853-100ul
100 μg
£500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18638303
View Documents
Novus Biologicals
NBP3-19853-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Casein Kinase 2 alpha Monoclonal antibody specifically detects Casein Kinase 2 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Gene Knock-Out
Specifications

Specifications

Casein Kinase 2 alpha
0.3 mg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
casein kinase 2, alpha 1 polypeptide, casein kinase II alpha 1 subunit, CK II alpha, CK2 catalytic subunit alpha, CK2A1casein kinase II subunit alpha, CKII, EC 2.7.11, EC 2.7.11.1, protein kinase CK2
A synthetic peptide of human Casein Kinase 2 alpha (Uniprot # P68400)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S05-7F8
Western Blot, Immunofluorescence, Immunoprecipitation, Gene Knock-Out
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, DNA Repair, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
1457
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.