missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 gamma Antibody, mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Casein Kinase 1 gamma Polyclonal antibody specifically detects Casein Kinase 1 gamma in Human samples. It is validated for ELISA
Specifications
Specifications
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Casein Kinase 1 gamma (Accession#: NP_071331.2; Met1-Lys422) |
| Quantity | 0.1 mL |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?