missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 epsilon Antibody (OTI5D4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70464PCP
This item is not returnable.
View return policy
Description
Casein Kinase 1 epsilon Monoclonal antibody specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Casein Kinase 1 epsilon | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI5D4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398 | |
| Human recombinant protein fragment corresponding to amino acids 153-416 of human CSNK1E (NP_689407) produced in E.coli. | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 1454 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction