missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 epsilon Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £392.00
Specifications
| Antigen | Casein Kinase 1 epsilon |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50-1:100, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676951
|
Novus Biologicals
NBP2-92968-0.02ml |
0.02 mL |
£186.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683770
|
Novus Biologicals
NBP2-92968-0.1ml |
0.1 mL |
£392.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Casein Kinase 1 epsilon Polyclonal antibody specifically detects Casein Kinase 1 epsilon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gene Knock-OutSpecifications
| Casein Kinase 1 epsilon | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Wnt Signaling Pathway | |
| PBS with 50% glycerol, pH7.3. | |
| 1454 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50-1:100, Immunohistochemistry-Paraffin, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| casein kinase 1, epsilon, casein kinase I isoform epsilon, CKIe, CKI-epsilon, EC 2.7.11, EC 2.7.11.1, HCKIE, MGC10398 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 247-416 of human Casein Kinase 1 epsilon (NP_689407.1). EFSTYLNFCRSLRFDDKPDYSYLRQLFRNLFHRQGFSYDYVFDWNMLKFGAARNPEDVDRERREHEREERMGQLRGSATRALPPGPPTGATANRLRSAAEPVASTPASRIQPAGNTSPRAISRVDRERKVSMRLHRGAPANVSSSDLTGRQEVSRIPASQTSVPFDHLGK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title