missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14388-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the casein kinase I (CKI) gene family whose members have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| Casein Kinase 1 delta | |
| Polyclonal | |
| Unconjugated | |
| CSNK1D | |
| Casein Kinase 1 delta, casein kinase 1, delta, Casein kinase I isoform delta, CKI delta, CKId, CSNK1D, HCKID | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104318, 1453, 64462 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P48730, Q06486, Q9DC28 | |
| Csnk1d | |
| Casein Kinase 1 delta Fusion Protein Ag5856 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction