missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 delta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52975
This item is not returnable.
View return policy
Description
Casein Kinase 1 delta Polyclonal specifically detects Casein Kinase 1 delta in Human samples. It is validated for Western Blot.
Specifications
| Casein Kinase 1 delta | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| casein kinase 1, delta, casein kinase I isoform delta, CKId, CKI-delta, EC 2.7.11, EC 2.7.11.1, HCKID | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| Protein Kinase | |
| 1453 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P48730 | |
| CSNK1D | |
| Synthetic peptides corresponding to CSNK1D(casein kinase 1, delta) The peptide sequence was selected from the middle region of CSNK1D. Peptide sequence NLVYIIDFGLAKKYRDARTHQHIPYRENKNLTGTARYASINTHLGIEQSR. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction