missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 alpha Antibody (2E2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30165C
This item is not returnable.
View return policy
Description
Casein Kinase 1 alpha Monoclonal antibody specifically detects Casein Kinase 1 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Casein Kinase 1 alpha | |
| Monoclonal | |
| DyLight 650 | |
| NP_001883 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 2E2 | |
| 50mM Sodium Borate | |
| casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975 | |
| Recombinant human Casein Kinase 1 alpha (1-337aa) purified from E. coli | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 1452 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction