missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Antibody, Novus Biologicals™
Description
Casein Polyclonal antibody specifically detects Casein in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Casein |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-500, ELISA 1:500-1000, Immunohistochemistry 1:100-500, Immunofluorescence, Immunohistochemistry-Paraffin 1:100-500 |
| Formulation | PBS |
| Gene Alias | alpha, alpha-S1-casein, CASA, casein alpha s1, MGC149368 |
| Host Species | Rabbit |
| Immunogen | The immunogen used to generate this antibody corresponds to bovine Casein. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?