missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASC5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92855-0.02ml
This item is not returnable.
View return policy
Description
CASC5 Polyclonal antibody specifically detects CASC5 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CASC5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| AF15Q14, Blinkin, blinkin, bub-linking kinetochore protein, Bub-linking kinetochore protein, cancer susceptibility candidate 5, Cancer susceptibility candidate gene 5 protein, Cancer/testis antigen 29, CT29KIAA1570, D40, hKNL-1, hSpc105, kinetochore null 1 homolog, Kinetochore-null protein 1, KNL1, KNL1ALL1-fused gene from chromosome 15q14 protein, protein CASC5, Protein D40/AF15q14 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CASC5 (NP_733468.3). MDGVSSEANEENDNIERPVRRRHSSILKPPRSPLQDLRGGNERVQESNALRNKKNSRRVSFADTIKVFQTESHMKIVRKSEMEGCSAMVPSQLQLLPPGFKRFSCLSLPETETGENLLLIQNKKLEDNYCEITGMNTLLSAPIHTQMQQKEFSIIEHTRERKHANDQTVIFSDENQMDLTSSHTVMITKGLLDNPISEKSTKIDTTSFLANLKLHTEDSRMKKEVNFSVDQNTSSENKIDFNDFIKRLKTGKCSAFPDVPDKENFEIPIYSKEPNSASSTHQMHVSLKEDENNSNITRLF | |
| 0.02 mL | |
| DNA Repair | |
| 57082 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction