missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CASC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30452
This item is not returnable.
View return policy
Description
CASC3 Polyclonal antibody specifically detects CASC3 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| CASC3 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| barentsz, Btz, cancer susceptibility candidate 3, Cancer susceptibility candidate gene 3 protein, metastatic lymph node 51, Metastatic lymph node gene 51 protein, MLN 51, MLN51BTZ, Protein barentsz, protein CASC3 | |
| between 653 and 703 | |
| 20 μg | |
| Cancer | |
| 22794 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction