missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | CASC3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CASC3 Polyclonal specifically detects CASC3 in Human samples. It is validated for Western Blot.Specifications
| CASC3 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| barentsz, Btz, cancer susceptibility candidate 3, Cancer susceptibility candidate gene 3 protein, metastatic lymph node 51, Metastatic lymph node gene 51 protein, MLN 51, MLN51BTZ, Protein barentsz, protein CASC3 | |
| CASC3 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| O15234 | |
| 22794 | |
| Synthetic peptides corresponding to CASC3(cancer susceptibility candidate 3) The peptide sequence was selected from the N terminal of CASC3. Peptide sequence MADRRRQRASQDTEDEESGASGSDSGGSPLRGGGSCSGSAGGGGSGSLPS. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title