missing translation for 'onlineSavingsMsg'
Learn More

CASC3 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18407334 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407334 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407334 Supplier Novus Biologicals Supplier No. NBP130321AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CASC3 Polyclonal antibody specifically detects CASC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASC3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias barentsz, Btz, cancer susceptibility candidate 3, Cancer susceptibility candidate gene 3 protein, metastatic lymph node 51, Metastatic lymph node gene 51 protein, MLN 51, MLN51BTZ, Protein barentsz, protein CASC3
Host Species Rabbit
Immunogen Maps to a region between residue 225 and 275 of human cancer susceptibility candidate 3 using the numbering given in entry NP_031385.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 22794
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.