Learn More
Thermo Scientific™ Cary-Blair with Indicator Single Vial
Product Code. 15703048
Change view
Click to view available options
Quantity:
12 Vials/Pk.
Closure Color:
Green
Unit Size:
Pack of 12
Product Code. Quantity Closure Color Unit Size
15703048 12 Vials/Pk. Green Pack of 12
1 options
Product Code. 15703048 Supplier Thermo Scientific™ Supplier No. R21610
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Collect, preserve, and recover enteric bacterial pathogens with a fecal transport that comes with industry color-coding and sporked caps and is stored at room temperature.
Collect, preserve, and recover enteric bacterial pathogens in stool specimens with Thermo Scientfic™ Cary-Blair with Indicator Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Maintain the viability of enteric bacterial pathogens during transport and storage with a formulation that includes 1.16% agar and is ready for use without any special preparation. Save valuable storage space with a transport that is stored in its original container at room temperature.
  • Cary-Blair with Indicator is an isotonic and nonnutritive medium, which does not contain a fermentable carbohydrate.
  • Sodium thioglycollate is added to impede oxidation.
  • Disodium phosphate is incorporated in the medium to act as a buffering agent, which prevents overgrowth of Escherichia coli and Enterobacter species.
  • The relatively high pH minimizes the destruction of bacteria due to acid formation.
  • Phenol red, a pH indicator, is added to demonstrate acidic shifts that can affect the viability of some enteric pathogens.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.
Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fecal Transport Vial
Closure Color Green
Description Cary-Blair with Indicator Single Vial (Green Cap)
Fill Volume 15 mL
Media Type Cary-Blair
Packaging Single vial. Packaged in partitioned boxes.
Quantity 12 Vials/Pk.