missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carveol(Mixture of cis and trans), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | (-)-Carveol |
| CAS | 99-48-9 |
| Molecular Formula | C10 H16 O |
| Quantity | 5 g |
| Synonym | 2-Cyclohexen-1-ol, 2-methyl-5-(1-methylethenyl)- (9CI, ACI), 2-Methyl-5-(1-methylethenyl)-2-cyclohexen-1-ol (ACI), p-Mentha-6,8-dien-2-ol (8CI), 2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-ol, 2-Methyl-5-isopropenyl-2-cyclohexen-1-ol, 2-Methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol, 5-Isopropenyl-2-methyl-2-cyclohexen-1-ol, 5-Isopropenyl-2-methylcyclohex-2-enol, Carveol, NSC 68313, p-Mentha-1,8-dien-6-ol |
| InChI Formula | InChI=1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9-11H,1,5-6H2,2-3H3 |
| SMILES | CC(=C)C1CC=C(C)C(O)C1 |
| IUPAC Name | 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol |
| Molecular Weight (g/mol) | 152.23 |
| Formula Weight | 152.1201 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction