missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carvedilol Phosphate Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Carvedilol phosphate hemihydrate |
| CAS | 610309-89-2 |
| Molecular Formula | 2 C24 H26 N2 O4 . H2 O . 2 H3 O4 P |
| Quantity | 500 mg |
| Synonym | 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, phosphate, hydrate (2:2:1), 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, phosphate (salt), hydrate (2:2:1) (9CI), Carvedilol dihydrogen phosphate hemihydrate, Carvedilol phosphate, Carvedilol phosphate hemihydrate, Coreg CR, SKF 105517D, SKF-105517D, Carvedilol phosphate hemihydrate, Carvedilol phosphate, SKF-105517D, SB-508446 |
| InChI Formula | InChI=1S/2C24H26N2O4.2H3O4P.H2O/c2*1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20;2*1-5(2,3)4;/h2*2-12,17,25-27H,13-16H2,1H3;2*(H3,1,2,3,4);1H2 |
| SMILES | O.COc1ccccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23.COc5ccccc5OCCNCC(O)COc6cccc7[nH]c8ccccc8c67.OP(=O)(O)O.OP(=O)(O)O |
| IUPAC Name | 1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol;phosphoric acid;hydrate |
| Molecular Weight (g/mol) | 1026.95 |
| Formula Weight | 1026.3429 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?