missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol Phosphate Hydrate, TRC

Product Code. 30390358
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390358 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390358 Supplier Toronto Research Chemicals Supplier No. TRCH279645100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carvedilol phosphate hemihydrate
CAS 610309-89-2
Molecular Formula 2 C24 H26 N2 O4 . H2 O . 2 H3 O4 P
Quantity 100 mg
Synonym 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, phosphate, hydrate (2:2:1), 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, phosphate (salt), hydrate (2:2:1) (9CI), Carvedilol dihydrogen phosphate hemihydrate, Carvedilol phosphate, Carvedilol phosphate hemihydrate, Coreg CR, SKF 105517D, SKF-105517D, Carvedilol phosphate hemihydrate, Carvedilol phosphate, SKF-105517D, SB-508446
InChI Formula InChI=1S/2C24H26N2O4.2H3O4P.H2O/c2*1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20;2*1-5(2,3)4;/h2*2-12,17,25-27H,13-16H2,1H3;2*(H3,1,2,3,4);1H2
SMILES O.COc1ccccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23.COc5ccccc5OCCNCC(O)COc6cccc7[nH]c8ccccc8c67.OP(=O)(O)O.OP(=O)(O)O
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol;phosphoric acid;hydrate
Molecular Weight (g/mol) 1026.95
Formula Weight 1026.3429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.