missing translation for 'onlineSavingsMsg'
Learn More

Carteolol hydrochloride, MedChemExpress

Product Code. 16403397
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16403397

Brand: Medchem Express HY17495A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma.

Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma.

Specifications

Chemical Name or Material Carteolol hydrochloride
CAS 51781-21-6
Color White
Purity Grade Notes Research
Molecular Formula C16H25ClN2O3
Quantity 50 mg
Synonym OPC-1085 hydrochloride
Solubility Information DMSO : 50 mg/mL (152.05 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1NC2=C(C(OCC(O)CNC(C)(C)C)=CC=C2)CC1.Cl
Molecular Weight (g/mol) 328.83
Formula Weight 328.83
Percent Purity 99.61%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.