missing translation for 'onlineSavingsMsg'
Learn More

Carteolol hydrochloride, MedChemExpress

Product Code. 16427336
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16427336 100 mg 100mg
16403397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16427336 Supplier Medchem Express Supplier No. HY17495A
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma.

Carteolol hydrochloride (OPC-1085 hydrochloride) is a non-selective beta blocker used to treat glaucoma.

Specifications

Chemical Name or Material Carteolol hydrochloride
CAS 51781-21-6
Color White
Purity Grade Notes Research
Molecular Formula C16H25ClN2O3
Quantity 100 mg
Synonym OPC-1085 hydrochloride
Solubility Information DMSO : 50 mg/mL (152.05 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1NC2=C(C(OCC(O)CNC(C)(C)C)=CC=C2)CC1.Cl
Molecular Weight (g/mol) 328.83
Formula Weight 328.83
Percent Purity 99.61%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.