missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carteolol hydrochloride

Product Code. 16564404 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16564404

Brand: Thermo Scientific Chemicals 464320050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51781-21-6
Molecular Formula C16H25ClN2O3
Molecular Weight (g/mol) 328.84
MDL Number MFCD00941499
InChI Key FYBXRCFPOTXTJF-UHFFFAOYNA-N
IUPAC Name hydrogen 5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydroquinolin-2-one chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)COC1=CC=CC2=C1CCC(=O)N2

Specifications

Melting Point 278.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: moderately soluble in methanol. slightly soluble, in acetic acid. insoluble in chloroform, ether,, dichloromethane
Formula Weight 328.83
Chemical Name or Material Carteolol hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.