missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CART/CARTPT Antibody (MM0124-7C11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12069AF405
This item is not returnable.
View return policy
Description
CART/CARTPT Monoclonal specifically detects CART/CARTPT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CART/CARTPT | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CARTPT | |
| Human recombinant CART/CARTPT | |
| 0.1 mL | |
| Primary | |
| Detects human CART/CARTPT protein (full length (aa28-116) and truncated aa69-116) | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MM0124-7C11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CART prepropeptide, CARTcocaine and amphetamine regulated transcript, cocaine- and amphetamine-regulated transcript protein | |
| Mouse | |
| Protein G purified | |
| Neuroscience | |
| 9607 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction