missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CARS Monoclonal Antibody (CL2310)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA524637

Product Code. 15729290

  • £480.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SQCNLYMAAR KAVRKRPNQA LLENIALYLT HMLKIFGAVE EDSSLGFPVG GPGTSLSLEA TVMPYLQVLS EFREGVRKIA REQKVPEILQ LSDALRDNIL PELGVRFEDH EGLPTVVKLV DRNTLLKERE EKRRVE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%. Binds to an epitope located within the peptide sequence EDSSLGFPVG as determined by overlapping synthetic peptides.

This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CARS
Monoclonal
1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P49589
CARS, CARS1
Recombinant protein fragment of Human CARS (Product #RP-101867)
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
CL2310
Unconjugated
CARS1
CA3; CARS; cars protein; CARS1; CYSRS; cysteine translase; cysteine tRNA ligase 1, cytoplasmic; cysteine--tRNA ligase, cytoplasmic; Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase 1; cysteinyl-tRNA synthetase, cytoplasmic; LOW QUALITY PROTEIN: cysteine--tRNA ligase, cytoplasmic; MGC:11246; sb:cb71
Mouse
Protein A
RUO
833
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.